4-Amino-3-methylbenzenesulfonic acid 98%

Supplier: Ambeed
Warning

A841581-1000G A841581-100G A841581-25G A841581-500G
76832-542EA 154.9 USD
76832-542 76832-544 76832-546 76832-548
4-Amino-3-methylbenzenesulfonic acid 98%
4-Amino-3-methylbenzenesulfonic acid

Precaución: for Research Use Only

Formula: C₇H₉NO₃S
MW: 187.22 g/mol
MDL Number: MFCD00025198
CAS Number: 98-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR