4-(6-Methoxybenzo[d]thiazol-2-yl)-N,N-dimethylaniline 95%

Supplier: Ambeed
Warning

A841468-100MG A841468-250MG
76832-536EA 85.94 USD
76832-536 76832-538
4-(6-Methoxybenzo[d]thiazol-2-yl)-N,N-dimethylaniline 95%
4-(6-Methoxybenzo[d]thiazol-2-yl)-N,N-dimethylaniline

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₂OS
MW: 284.38 g/mol
CAS Number: 10205-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR