N-Boc-2-(5-bromo-2-thienyl)ethanamine 97%
Supplier: Ambeed
|
Warning
|
A836218-1G
A836218-10G
76832-528EA
119.89
USD
76832-528
77684-796
N-Boc-2-(5-bromo-2-thienyl)ethanamine 97%
N-Boc-2-(5-bromo-2-thienyl)ethanamine
Precaución: For research use only.
Formula:
C₁₁H₁₆BrNO₂S MW: 306.21 g/mol |
MDL Number:
MFCD07371534 CAS Number: 885279-60-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...