(3-((2-Methoxyethoxy)methyl)phenyl)boronic acid 96%
Supplier: Ambeed
|
Warning
|
A834564-100MG
A834564-1G
A834564-250MG
76832-522EA
45.62
USD
76832-522
76832-524
76832-526
(3-((2-Methoxyethoxy)methyl)phenyl)boronic acid 96%
(3-((2-Methoxyethoxy)methyl)phenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₅BO₄ MW: 210.03 g/mol |
MDL Number:
MFCD14687274 CAS Number: 1256358-61-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...