4-Methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 5-Cyano-2-methoxyphenylboronic acid pinacol ester

A834019-1G A834019-250MG A834019-5G
76832-516EA 56.23 USD
76832-516 76832-518 76832-520
4-Methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 97%
4-Methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₈BNO₃
MW: 259.11 g/mol
MDL Number: MFCD10700153
CAS Number: 706820-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR