4,4,5,5-Tetramethyl-2-(4-methylnaphthalen-1-yl)-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

A833662-100MG A833662-1G A833662-250MG A833662-5G
76832-508EA 27.59 USD
76832-508 76832-510 76832-512 76832-514
4,4,5,5-Tetramethyl-2-(4-methylnaphthalen-1-yl)-1,3,2-dioxaborolane 98%
4,4,5,5-Tetramethyl-2-(4-methylnaphthalen-1-yl)-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₇H₂₁BO₂
MW: 268.16 g/mol
MDL Number: MFCD20231463
CAS Number: 627526-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR