1,3-Dimethyl-8-nitro-1H-purine-2,6(3H,9H)-dione 95+%

Supplier: Ambeed
Warning

A830614-1G A830614-250MG A830614-5G
76832-482EA 60.48 USD
76832-482 76832-484 76832-486
1,3-Dimethyl-8-nitro-1H-purine-2,6(3H,9H)-dione 95+%
1,3-Dimethyl-8-nitro-1H-purine-2,6(3H,9H)-dione

Precaución: for Research Use Only

Formula: C₇H₇N₅O₄
MW: 225.16 g/mol
MDL Number: MFCD09400563
CAS Number: 2099-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR