(R)-3,3'-Bis(2,4,6-triisopropylphenyl)-[1,1'-binapthalene]-2,2'-diol 97%

Supplier: Ambeed
Warning

Synonyms: (R)-3,3'-Bis(2,4,6-triisopropylphenyl)-[1,1'-binaphthalene]-2,2'-diol

A829680-100MG A829680-1G A829680-250MG
76832-476EA 211.12 USD
76832-476 76832-478 76832-480
(R)-3,3'-Bis(2,4,6-triisopropylphenyl)-[1,1'-binapthalene]-2,2'-diol 97%
(R)-3,3'-Bis(2,4,6-triisopropylphenyl)-[1,1'-binapthalene]-2,2'-diol

Precaución: for Research Use Only

Formula: C₅₀H₅₈O₂
MW: 690.994 g/mol
MDL Number: MFCD11045378
CAS Number: 247123-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR