3,4-Dichlorothiophene-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A829528-1G A829528-250MG
76832-466EA 774.46 USD
76832-466 76832-468
3,4-Dichlorothiophene-2-carboxylic acid 98%
3,4-Dichlorothiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₂Cl₂O₂S
MW: 197.03 g/mol
MDL Number: MFCD00159568
CAS Number: 61209-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR