1,2,3-Trimethyl-3H-benzo[e]indole 98%

Supplier: Ambeed
Warning

A822639-1G A822639-250MG A822639-5G
76832-450EA 26 USD
76832-450 76832-452 76832-454
1,2,3-Trimethyl-3H-benzo[e]indole 98%
1,2,3-Trimethyl-3H-benzo[e]indole

Precaución: for Research Use Only

Formula: C₁₅H₁₅N
MDL Number: MFCD06651515
CAS Number: 881219-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR