3-Ethoxy-4-(3-methoxypropoxy)benzaldehyde 97%

Supplier: Ambeed
Warning

A822544-1G A822544-250MG A822544-5G
76832-444EA 28 USD
76832-444 76832-446 76832-448
3-Ethoxy-4-(3-methoxypropoxy)benzaldehyde 97%
3-Ethoxy-4-(3-methoxypropoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₄
MDL Number: MFCD13204852
CAS Number: 946779-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR