(R)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-bi-2-napthol 98%

Supplier: Ambeed
Warning

Synonyms: (R)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-[1,1'-binaphthalene]-2,2'-diol, (R)-3,3'-Bis[3,5-bis(trifluoromethyl)phenyl]1,1'-binaphthalene-2,2'-diol

A822395-100MG A822395-1G A822395-250MG A822395-50MG A822395-5G
76832-434EA 39 USD
76832-434 76832-436 76832-438 76832-440 76832-442
(R)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-bi-2-napthol 98%
(R)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-bi-2-napthol

Precaución: for Research Use Only

Formula: C₃₆H₁₈F₁₂O₂
MDL Number: MFCD08689862
CAS Number: 756491-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR