5-Chloro-1H-indol-6-amine 98%

Supplier: Ambeed
Warning

A822278-100MG A822278-1G A822278-250MG
76832-428EA 131.56 USD
76832-428 76832-430 76832-432
5-Chloro-1H-indol-6-amine 98%
5-Chloro-1H-indol-6-amine

Precaución: for Research Use Only

Formula: C₈H₇ClN₂
MW: 166.60 g/mol
MDL Number: MFCD12924460
CAS Number: 873055-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR