3-Methylisothiazol-5-amine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-3-methyl-isothiazole hydrochloride, 5-Amino-3-methylisothiazole hydrochloride

A820340-100G A820340-10G A820340-1G A820340-250MG A820340-25G A820340-5G
76832-410EA 627 USD
76832-410 76832-412 76832-414 76832-416 76832-418 76832-420
3-Methylisothiazol-5-amine hydrochloride 97%
3-Methylisothiazol-5-amine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₇ClN₂S
MW: 150.63 g/mol
MDL Number: MFCD00012716
CAS Number: 52547-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR