3-Chloro-5-(trifluoromethoxy)benzaldehyde 98%

Supplier: Ambeed
Warning

A814151-1G A814151-250MG
76832-398EA 53.05 USD
76832-398 76832-400
3-Chloro-5-(trifluoromethoxy)benzaldehyde 98%
3-Chloro-5-(trifluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₂
MDL Number: MFCD06660305
CAS Number: 433926-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR