5-(tert-Butyl)pyridin-2-amine 95%

Supplier: Ambeed
Warning

A810933-100MG A810933-1G A810933-250MG A810933-50MG
76832-380EA 204.76 USD
76832-380 76832-382 76832-384 76832-386
5-(tert-Butyl)pyridin-2-amine 95%
5-(tert-Butyl)pyridin-2-amine

Precaución: for Research Use Only

Formula: C₉H₁₄N₂
MW: 150.22 g/mol
MDL Number: MFCD12033220
CAS Number: 866620-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR