5-Chloro-2-methylnicotinaldehyde 97%

Supplier: Ambeed
Warning

A809016-100MG A809016-250MG
76832-366EA 89.12 USD
76832-366 76832-368
5-Chloro-2-methylnicotinaldehyde 97%
5-Chloro-2-methylnicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₆ClNO
MW: 155.58 g/mol
MDL Number: MFCD10698573
CAS Number: 1060814-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR