Z-D-Ser-OMe 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: N-Cbz-D-serine Methyl Ester, Cbz-D-Ser-OMe

A806879-100G A806879-25G A806879-5G A806879-500G
76832-360EA 107 USD
76832-360 76832-362 76832-364 77684-755
Z-D-Ser-OMe 98%, Ambeed.Inc
Z-D-Ser-OMe

Precaución: For research use only.

Formula: C₁₂H₁₅NO₅
MW: 253.2 g/mol
MDL Number: MFCD00799501
CAS Number: 93204-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR