(R)-(S)-Josiphos 97%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: (2R)-1-[(1R)-1-(Dicyclohexylphosphino)ethyl]-2-(diphenylphosphino)ferrocene, (2Sp)-1-[(1R)-1-(Dicyclohexylphosphino)ethyl]-2-(diphenylphosphino)ferrocene, (R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine ethanol adduct, (R)-(-)-1-[(S)-2-Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine

A800274-100MG A800274-1G A800274-250MG A800274-25G A800274-5G
76832-324EA 6 USD
76832-324 76832-326 76832-328 77684-744 77684-745
(R)-(S)-Josiphos 97%, Ambeed.Inc
(R)-(S)-Josiphos

Precaución: For research use only.

Formula: C₃₆H₄₄FeP₂
MW: 594.541 g/mol
MDL Number: MFCD00800284
CAS Number: 155806-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR