Methyl 5-nitro-2-(trifluoromethyl)benzoate 97%

Supplier: Ambeed
Warning

A800114-100MG A800114-250MG
76832-320EA 59.42 USD
76832-320 76832-322
Methyl 5-nitro-2-(trifluoromethyl)benzoate 97%
Methyl 5-nitro-2-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₆F₃NO₄
MW: 249.14 g/mol
MDL Number: MFCD14698084
CAS Number: 1214328-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR