2-Pyrrolidino-5-pyrimidineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Pyrrolidin-1-yl)pyrimidine-5-boronic acid pinacol ester, 2-(Pyrrolidin-1-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

A799920-250MG A799920-1G
76832-318EA 61.54 USD
76832-318 77684-743
2-Pyrrolidino-5-pyrimidineboronic acid pinacol ester 98%
2-Pyrrolidino-5-pyrimidineboronic acid pinacol ester

Precaución: For research use only.

Formula: C₁₄H₂₂BN₃O₂
MW: 275.1544 g/mol
MDL Number: MFCD07781249
CAS Number: 1015242-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR