Methyl 5-bromo-6-methylpicolinate 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-6-methylpyridine-2-carboxylic acid methyl ester

A799210-100MG A799210-10G A799210-1G A799210-250MG A799210-5G
76832-308EA 65.78 USD
76832-308 76832-310 76832-312 76832-314 76832-316
Methyl 5-bromo-6-methylpicolinate 97%
Methyl 5-bromo-6-methylpicolinate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.06 g/mol
MDL Number: MFCD09753045
CAS Number: 1215860-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR