(R)-4-Boc-3-(aminomethyl)-morpholine 98%

Supplier: Ambeed
Warning

Synonyms: (R)-tert-Butyl 3-(aminomethyl)morpholine-4-carboxylate

A797881-50MG A797881-100MG A797881-1G
76832-298EA 40 USD
76832-298 77886-777 77886-778
(R)-4-Boc-3-(aminomethyl)-morpholine 98%
(R)-4-Boc-3-(aminomethyl)-morpholine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₃
MW: 216.277 g/mol
MDL Number: MFCD06739119
CAS Number: 1187929-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR