3,6-Dihydroxy-9H-xanthen-9-one 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A796011-100MG A796011-1G A796011-250MG A796011-25G A796011-5G
76832-280EA 15 USD
76832-280 76832-282 76832-284 77684-739 77684-740
3,6-Dihydroxy-9H-xanthen-9-one 95%, Ambeed.Inc
3,6-Dihydroxy-9H-xanthen-9-one

Precaución: For research use only.

Formula: C₁₃H₈O₄
MDL Number: MFCD00226967
CAS Number: 1214-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR