2-Ethoxy-5-(trifluoromethyl)-3-pyridineboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (2-Ethoxy-5-(trifluoromethyl)pyridin-3-yl)boronic acid

A794321-1G A794321-250MG
76832-264EA 89.12 USD
76832-264 76832-266
2-Ethoxy-5-(trifluoromethyl)-3-pyridineboronic acid 95%
2-Ethoxy-5-(trifluoromethyl)-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BF₃NO₃
MW: 234.9682 g/mol
MDL Number: MFCD15143470
CAS Number: 1218790-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR