N-(4-Bromophenyl)benzo[d]thiazol-2-amine 95%

Supplier: Ambeed
Warning

A792953-100MG A792953-1G A792953-250MG
76832-258EA 14.86 USD
76832-258 76832-260 76832-262
N-(4-Bromophenyl)benzo[d]thiazol-2-amine 95%
N-(4-Bromophenyl)benzo[d]thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₃H₉BrN₂S
MW: 305.19 g/mol
MDL Number: MFCD03460423
CAS Number: 6278-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR