1-(2-Chloropropyl)piperidine 95%

Supplier: Ambeed
Warning

A791419-100MG A791419-1G A791419-250MG A791419-5G
76832-238EA 25.47 USD
76832-238 76832-240 76832-242 76832-244
1-(2-Chloropropyl)piperidine 95%
1-(2-Chloropropyl)piperidine

Precaución: for Research Use Only

Formula: C₈H₁₆ClN
MW: 161.67 g/mol
MDL Number: MFCD19105113
CAS Number: 698-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR