1-[(2-Trimethylsilylethoxy)methyl]-1H-pyrazole-5-boronic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(1-((2-(Trimethylsilyl)ethoxy)methyl)-1H-pyrazol-5-yl)boronic acid
A790961-100MG
A790961-250MG
A790961-1G
76832-234EA
35.01
USD
76832-234
76832-236
77684-733
1-[(2-Trimethylsilylethoxy)methyl]-1H-pyrazole-5-boronic acid 95%
1-[(2-Trimethylsilylethoxy)methyl]-1H-pyrazole-5-boronic acid
Precaución: For research use only.
Formula:
C₉H₁₉BN₂O₃Si MW: 242.1600 g/mol |
MDL Number:
MFCD11855854 CAS Number: 188978-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...