3-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 98%
Supplier: Ambeed
|
Warning
|
A790185-100MG
A790185-1G
A790185-250MG
A790185-5G
76832-228EA
11
USD
76832-228
76832-230
76832-232
77886-763
3-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 98%
3-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Precaución: for Research Use Only
Formula:
C₁₃H₁₅BFNO₂ |
MDL Number:
MFCD11521351 CAS Number: 935685-88-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...