4-Nitrophthalide 98%

Supplier: Ambeed
Warning

A789312-100MG A789312-1G A789312-250MG A789312-5G
76832-220EA 7 USD
76832-220 76832-222 76832-224 76832-226
4-Nitrophthalide 98%
4-Nitrophthalide

Precaución: for Research Use Only

Formula: C₈H₅NO₄
MDL Number: MFCD03069001
CAS Number: 65399-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR