Ethyl 6-bromo-2-chloronicotinate 95%

Supplier: Ambeed
Warning

A788926-100MG A788926-1G A788926-250MG
76832-210EA 99.73 USD
76832-210 76832-212 76832-214
Ethyl 6-bromo-2-chloronicotinate 95%
Ethyl 6-bromo-2-chloronicotinate

Precaución: for Research Use Only

Formula: C₈H₇BrClNO₂
MW: 264.50 g/mol
MDL Number: MFCD14698315
CAS Number: 1214335-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR