(S)-3,3'-Di(phenanthren-9-yl)-1,1'-binaphthyl-2,2'-diol 98%

Supplier: Ambeed
Warning

Synonyms: (S)-3,3'-Di(anthracen-9-yl)-[1,1'-binaphthalene]-2,2'-diol1

A786394-100MG A786394-250MG A786394-50MG A786394-1G
76832-188EA 76.39 USD
76832-188 76832-190 76832-192 77684-727
(S)-3,3'-Di(phenanthren-9-yl)-1,1'-binaphthyl-2,2'-diol 98%
(S)-3,3'-Di(phenanthren-9-yl)-1,1'-binaphthyl-2,2'-diol

Precaución: For research use only.

Formula: C₄₈H₃₀O₂
MW: 638.7500 g/mol
MDL Number: MFCD09027270
CAS Number: 361342-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR