Ethyl thieno[3,2-b]thiophene-2-carboxylate 95+%

Supplier: Ambeed
Warning

A785848-250MG A785848-5G
76832-178EA 28 USD
76832-178 76832-180
Ethyl thieno[3,2-b]thiophene-2-carboxylate 95+%
Ethyl thieno[3,2-b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈O₂S₂
MDL Number: MFCD06803452
CAS Number: 201004-08-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR