Methyl 5-cyclopropylpyrazole-4-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-cyclopropyl-1H-pyrazole-4-carboxylate

A785510-1G A785510-250MG
76832-174EA 60.48 USD
76832-174 76832-176
Methyl 5-cyclopropylpyrazole-4-carboxylate 98%
Methyl 5-cyclopropylpyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.177 g/mol
MDL Number: MFCD11855888
CAS Number: 1150164-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR