1-(Azetidin-3-yl)-1H-imidazole hydrochloride 95%

Supplier: Ambeed
Warning

A785260-1G A785260-250MG
76832-170EA 163 USD
76832-170 76832-172
1-(Azetidin-3-yl)-1H-imidazole hydrochloride 95%
1-(Azetidin-3-yl)-1H-imidazole hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₀ClN₃
MDL Number: MFCD22056235
CAS Number: 153836-44-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR