2-Chloro-6-(trifluoromethyl)benzyl bromide 95%

Supplier: Ambeed
Danger

A784273-1G A784273-250MG
76832-168EA 20 USD
76832-168 77886-757
2-Chloro-6-(trifluoromethyl)benzyl bromide 95%
2-Chloro-6-(trifluoromethyl)benzyl bromide

Precaución: for Research Use Only

Formula: C₈H₅BrClF₃
MDL Number: MFCD06660232
CAS Number: 886500-26-1
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR