6-Bromo-4,4-dimethylthiochroman 97%

Supplier: Ambeed
Warning

A784151-1G A784151-250MG
76832-162EA 31 USD
76832-162 76832-164
6-Bromo-4,4-dimethylthiochroman 97%
6-Bromo-4,4-dimethylthiochroman

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrS
MW: 257.18 g/mol
MDL Number: MFCD15144704
CAS Number: 112110-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR