tert-Butyl 3,9-diazaspiro[5.5]undecane-3-carboxylate hydrochloride 97%

Supplier: Ambeed
Warning

A783079-100MG A783079-250MG A783079-10G A783079-1G A783079-5G
76832-158EA 24.41 USD
76832-158 76832-160 77614-392 77614-394 77614-396
tert-Butyl 3,9-diazaspiro[5.5]undecane-3-carboxylate hydrochloride 97%
tert-Butyl 3,9-diazaspiro[5.5]undecane-3-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₂₇ClN₂O₂
MW: 290.82 g/mol
MDL Number: MFCD11656757
CAS Number: 236406-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR