(2R,6R)-2,6-Dimethylmorphline 98%

Supplier: Ambeed
Danger

Synonyms: (2R,6R)-2,6-Dimethylmorpholine

A782107-1G A782107-100MG A782107-250MG
76832-154EA 375.56 USD
76832-154 77684-721 77684-722
(2R,6R)-2,6-Dimethylmorphline 98%
(2R,6R)-2,6-Dimethylmorphline

Precaución: For research use only.

Formula: C₆H₁₃NO
MW: 115.174 g/mol
MDL Number: MFCD18250270
CAS Number: 171753-74-5
UN: 1992
ADR: 3,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR