(3-((Neopentyloxy)sulfonyl)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A782059-100MG A782059-1G A782059-250MG
76832-148EA 36.08 USD
76832-148 76832-150 76832-152
(3-((Neopentyloxy)sulfonyl)phenyl)boronic acid 98%
(3-((Neopentyloxy)sulfonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇BO₅S
MW: 272.13 g/mol
MDL Number: MFCD12025962
CAS Number: 951233-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR