4,4,5,5-Tetramethyl-2-(4-(methylsulfinyl)phenyl)-1,3,2-dioxaborolane 95%
Supplier: Ambeed
|
Danger
|
A781294-100MG
A781294-1G
A781294-250MG
A781294-5G
76832-140EA
39
USD
76832-140
76832-142
76832-144
76832-146
4,4,5,5-Tetramethyl-2-(4-(methylsulfinyl)phenyl)-1,3,2-dioxaborolane 95%
4,4,5,5-Tetramethyl-2-(4-(methylsulfinyl)phenyl)-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₁₃H₁₉BO₃S |
MDL Number:
MFCD16660301 CAS Number: 1016641-70-5 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...