(4-Butyrylphenyl)boronic acid 98%

Supplier: Ambeed
Warning

A778176-1G A778176-250MG A778176-5G
76832-130EA 207 USD
76832-130 76832-132 76832-134
(4-Butyrylphenyl)boronic acid 98%
(4-Butyrylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BO₃
MDL Number: MFCD08701778
CAS Number: 186498-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR