9-Benzyl-2,6-dichloro-9H-purine 95%

Supplier: Ambeed
Warning

A777380-10G A777380-1G A777380-250MG A777380-5G
76832-116EA 115 USD
76832-116 76832-118 76832-120 76832-122
9-Benzyl-2,6-dichloro-9H-purine 95%
9-Benzyl-2,6-dichloro-9H-purine

Precaución: for Research Use Only

Formula: C₁₂H₈Cl₂N₄
MDL Number: MFCD11047303
CAS Number: 79064-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR