3,6-Dichloropyridazine-4-carboxamide 98%

Supplier: Ambeed
Warning

A775274-250MG A775274-1G
76832-104EA 128.37 USD
76832-104 77684-715
3,6-Dichloropyridazine-4-carboxamide 98%
3,6-Dichloropyridazine-4-carboxamide

Precaución: For research use only.

Formula: C₅H₃Cl₂N₃O
MW: 192.00 g/mol
MDL Number: MFCD11646064
CAS Number: 27427-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR