5-Nitro-2H-3,1-benzoxazine-2,4(1H)-dione 95%

Supplier: Ambeed
Warning

Synonyms: 5-Nitro-1H-benzo[d][1,3]oxazine-2,4-dione

A775157-100MG A775157-1G A775157-250MG A775157-5G
76832-096EA 31.83 USD
76832-096 76832-098 76832-100 76832-102
5-Nitro-2H-3,1-benzoxazine-2,4(1H)-dione 95%
5-Nitro-2H-3,1-benzoxazine-2,4(1H)-dione

Precaución: for Research Use Only

Formula: C₈H₄N₂O₅
MW: 208.13 g/mol
MDL Number: MFCD00016919
CAS Number: 20829-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR