(4R,4'R)-2,2'-(Propane-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole) ≥98%, ee 99%

Supplier: Ambeed
Warning

A774579-100MG A774579-1G A774579-250MG A774579-5G
76832-092EA 24.41 USD
76832-092 76832-094 77614-378 77684-714
(4R,4'R)-2,2'-(Propane-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole) ≥98%, ee 99%
(4R,4'R)-2,2'-(Propane-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole)

Precaución: For research use only.

Formula: C₂₃H₂₆N₂O₂
MW: 362.46 g/mol
MDL Number: MFCD01863573
CAS Number: 141362-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR