1-Acryloyloxy-3-hydroxyadamantane 98%

Supplier: Ambeed
Warning

Synonyms: Acrylic acid 3-Hydroxy-1-adamantyl ester, 3-Hydroxyadamantan-1-yl acrylate, 3-Hydroxy-1-adamantyl acrylate

A774573-1G A774573-25G A774573-5G
76832-086EA 14.86 USD
76832-086 76832-088 76832-090
1-Acryloyloxy-3-hydroxyadamantane 98%
1-Acryloyloxy-3-hydroxyadamantane

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
MW: 222.28 g/mol
MDL Number: MFCD13194929
CAS Number: 216581-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR