4-Chloro-2-fluorobenzamide 95%

Supplier: Ambeed
Warning

A772223-1G A772223-25G A772223-5G A772223-100G
76832-074EA 7.43 USD
76832-074 76832-076 76832-078 77684-712
4-Chloro-2-fluorobenzamide 95%
4-Chloro-2-fluorobenzamide

Precaución: For research use only.

Formula: C₇H₅ClFNO
MW: 173.572 g/mol
MDL Number: MFCD04115956
CAS Number: 104326-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR