6-(3,4-Diaminophenyl)-5-methyl-4,5-dihydropyridazin-3(2H)-one 97%

Supplier: Ambeed
Warning

A768535-250MG A768535-1G
76832-068EA 12 USD
76832-068 77886-738
6-(3,4-Diaminophenyl)-5-methyl-4,5-dihydropyridazin-3(2H)-one 97%
6-(3,4-Diaminophenyl)-5-methyl-4,5-dihydropyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₄O
MDL Number: MFCD00761653
CAS Number: 74150-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR