(4,6-Dimethylpyridin-2-yl)methanol 95%

Supplier: Ambeed
Warning

A767841-1G A767841-250MG
76832-066EA 279.02 USD
76832-066 77684-705
(4,6-Dimethylpyridin-2-yl)methanol 95%
(4,6-Dimethylpyridin-2-yl)methanol

Precaución: For research use only.

Formula: C₈H₁₁NO
MW: 137.17 g/mol
MDL Number: MFCD00956273
CAS Number: 18087-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR